Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrriq4
Entrez Gene ID
68307
Gene Name
leucine-rich repeats and IQ motif containing 4
Gene Aliases
4930558O21Rik
Chromosome Location
Chr.3: 30644506 - 30672431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026668.2 NP_080944.2 3 - 4 536 139
GenBank mRNA AK019775.1 - 3 - 4 537 139
BC145806.1 - 3 - 4 237 139

View Full Product Details