Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Igsf9
Entrez Gene ID
93842
Gene Name
immunoglobulin superfamily, member 9
Gene Aliases
644ETD8, Dasm1, Kiaa1355-hp, NRT1, Ncaml, mKIAA1355
Chromosome Location
Chr.1: 172481776 - 172498877 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145800.1 NP_001139272.1 5 - 6 806 75
NM_033608.3 NP_291086.2 5 - 6 872 75
GenBank mRNA AF317839.1 - 5 - 6 872 75
AK078536.1 - 5 - 6 800 75
AK145121.1 - 5 - 6 781 75
BC034594.1 - 5 - 6 788 75
BC040281.1 - 5 - 6 832 75

View Full Product Details