Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casq1
Entrez Gene ID
12372
Gene Name
calsequestrin 1
Gene Aliases
CSQ, CSQ-1, CSQ1, sCSQ
Chromosome Location
Chr.1: 172209894 - 172219895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009813.2 NP_033943.2 9 - 10 1193 84
GenBank mRNA AK049865.1 - 3 - 4 311 84
BC064057.1 - 9 - 10 1131 84
U93291.1 - 9 - 10 1149 84

View Full Product Details