The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029294.2 | NP_083570.1 | 1 - 1 | 1618 | 153 |
GenBank mRNA | AK077203.1 | - | 1 - 1 | 1619 | 153 |