The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_205769.2 | NP_991338.1 | 2 - 2 | 899 | 128 |
GenBank mRNA | BC116921.1 | - | 1 - 1 | 681 | 128 |
BC119036.1 | - | 1 - 1 | 681 | 128 |