The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206896.2 | NP_996779.1 | 1 - 1 | 994 | 60 |
GenBank mRNA | BC120559.1 | - | 1 - 1 | 994 | 60 |
BC120561.1 | - | 1 - 1 | 994 | 60 |