The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010766.2 | NP_034896.1 | 4 - 5 | 771 | 141 |
GenBank mRNA | AK156388.1 | - | 4 - 5 | 445 | 141 |
AK159967.1 | - | 4 - 5 | 453 | 141 | |
AK169230.1 | - | 4 - 5 | 447 | 141 | |
BC132338.1 | - | 4 - 5 | 711 | 141 | |
BC137939.1 | - | 4 - 5 | 711 | 141 | |
U18424.1 | - | 4 - 5 | 623 | 141 |