The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021433.3 | NP_067408.1 | 4 - 5 | 553 | 58 |
GenBank mRNA | AF237814.1 | - | 4 - 5 | 363 | 58 |
AK009690.1 | - | 4 - 5 | 508 | 58 | |
AK018167.1 | - | 4 - 5 | 541 | 58 | |
AK019106.1 | - | 4 - 5 | 515 | 58 | |
AK031673.1 | - | 4 - 5 | 532 | 58 | |
AK044905.1 | - | 4 - 5 | 514 | 58 | |
AK049124.1 | - | 4 - 5 | 513 | 58 | |
AK151549.1 | - | 4 - 5 | 507 | 58 | |
AK159392.1 | - | 4 - 5 | 525 | 58 | |
AK179436.1 | - | 3 - 4 | 201 | 58 | |
BC029205.1 | - | 4 - 5 | 470 | 58 | |
BQ749205.1 | - | 4 - 5 | 526 | 58 |