Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx16
Entrez Gene ID
74718
Gene Name
sorting nexin 16
Gene Aliases
4930522N22Rik, AI117577, AI747675
Chromosome Location
Chr.3: 10417817 - 10440131 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127191.2 NP_001120663.1 1 - 2 217 153
NM_029068.4 NP_083344.3 1 - 2 217 153
GenBank mRNA AK015872.1 - 1 - 2 178 153
AK032055.1 - 1 - 2 174 153
AK043975.1 - 1 - 2 187 153
AK166422.1 - 1 - 2 191 153
BC070431.1 - - 162 153

View Full Product Details