Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx16
Entrez Gene ID
74718
Gene Name
sorting nexin 16
Gene Aliases
4930522N22Rik, AI117577, AI747675
Chromosome Location
Chr.3: 10417817 - 10440131 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029068.4 NP_083344.3 6 - 7 1133 105
GenBank mRNA AK015872.1 - 6 - 7 1094 105
AK032055.1 - 6 - 7 1090 105
BC019424.1 - 5 - 6 898 105
BC070431.1 - - 1076 105

View Full Product Details