Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spp2
Entrez Gene ID
75396
Gene Name
secreted phosphoprotein 2
Gene Aliases
0610038O04Rik, pp-24, spp24
Chromosome Location
Chr.1: 88406967 - 88428300 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029269.2 NP_083545.1 7 - 8 752 90
GenBank mRNA AJ315513.1 - 7 - 8 713 90
AK002814.1 - 7 - 8 753 90
BC027494.1 - 7 - 8 678 90

View Full Product Details