The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019645.2 | NP_062619.1 | 13 - 14 | 2227 | 70 |
GenBank mRNA | AK036856.1 | - | 13 - 14 | 2412 | 70 |
AK079429.1 | - | 13 - 14 | 2412 | 70 | |
Y07941.1 | - | 13 - 14 | 2227 | 70 |