Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Casc3
Entrez Gene ID
192160
Gene Name
cancer susceptibility candidate 3
Gene Aliases
Btz, Mln51
Chromosome Location
Chr.11: 98809808 - 98833807 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138660.2 NP_619601.2 4 - 5 502 119
GenBank mRNA AF526276.1 - 4 - 5 476 119
AJ292072.1 - - 716 119
AK139608.1 - 4 - 5 541 119
AK145886.1 - 4 - 5 276 119
BC034533.1 - 1 - 2 146 119
BC060672.1 - 4 - 5 330 119
BC141296.1 - 4 - 5 615 119

View Full Product Details