Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Eme1
- Entrez Gene ID
-
268465
- Gene Name
- essential meiotic endonuclease 1 homolog 1 (S. pombe)
- Gene Aliases
-
6820428D13
- Chromosome Location
-
Chr.11: 94644998 - 94653785 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177752.4 |
NP_808420.1 |
1 - 2 |
59 |
75 |
View Full Product Details