The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013367.3 | NP_001013385.3 | 8 - 9 | 1441 | 139 |
GenBank mRNA | AK081874.1 | - | 2 - 3 | 296 | 139 |
AK147811.1 | - | 8 - 9 | 1419 | 139 | |
AK160612.1 | - | 8 - 9 | 1422 | 139 | |
AY885266.1 | - | 8 - 9 | 1400 | 139 | |
BC086695.1 | - | 8 - 9 | 1411 | 139 |