Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxf7
Entrez Gene ID
170722
Gene Name
nuclear RNA export factor 7
Gene Aliases
Nx7, Nxf6, nfx-a1, nxf-a, nxf-a2
Chromosome Location
Chr.X: 135579555 - 135598777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130888.1 NP_570958.1 15 - 16 1291 109
GenBank mRNA AB178901.1 - 16 - 17 1430 109
AJ305317.1 - 11 - 12 852 109
AJ305318.1 - 12 - 13 960 109
AY260550.1 - 15 - 16 1291 109

View Full Product Details