Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxf7
Entrez Gene ID
170722
Gene Name
nuclear RNA export factor 7
Gene Aliases
Nx7, Nxf6, nfx-a1, nxf-a, nxf-a2
Chromosome Location
Chr.X: 135579555 - 135598777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130888.1 NP_570958.1 9 - 10 802 63
GenBank mRNA AB178901.1 - 10 - 11 941 63
AJ305318.1 - 6 - 7 471 63
AY260550.1 - 9 - 10 802 63

View Full Product Details