Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrk2
Entrez Gene ID
66725
Gene Name
leucine-rich repeat kinase 2
Gene Aliases
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
Chromosome Location
Chr.15: 91672821 - 91816124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025730.3 NP_080006.3 48 - 49 7240 100
GenBank mRNA AK014886.1 - 9 - 10 1299 100
AK014938.1 - 9 - 10 1312 100
AK156100.1 - 1 - 2 2913 100
AY792512.1 - 48 - 49 7183 100
BC034074.1 - 14 - 15 2123 100
BC035949.1 - 4 - 5 584 100

View Full Product Details