Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrk2
Entrez Gene ID
66725
Gene Name
leucine-rich repeat kinase 2
Gene Aliases
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
Chromosome Location
Chr.15: 91672821 - 91816124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025730.3 NP_080006.3 36 - 37 5374 122
GenBank mRNA AK034413.1 - 13 - 14 1997 122
AY792512.1 - 36 - 37 5317 122
BC034074.1 - 2 - 3 257 122

View Full Product Details