Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccl8
Entrez Gene ID
20307
Gene Name
chemokine (C-C motif) ligand 8
Gene Aliases
1810063B20Rik, AB023418, HC14, MCP-2, Mcp2, Scya8
Chromosome Location
Chr.11: 82115185 - 82116799 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021443.3 NP_067418.1 2 - 3 238 84
GenBank mRNA AB023418.1 - 2 - 3 246 84
AK007942.1 - 2 - 3 239 84
BC117101.1 - 2 - 3 236 84
BC117103.1 - 2 - 3 236 84
BY707942.1 - 2 - 3 239 84

View Full Product Details