Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d22b
Entrez Gene ID
381085
Gene Name
TBC1 domain family, member 22B
Gene Aliases
BC045600
Chromosome Location
Chr.17: 29549788 - 29606894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198647.1 NP_941049.1 7 - 8 999 73
GenBank mRNA BC045600.1 - 7 - 8 999 73
BC060066.1 - 2 - 3 271 73

View Full Product Details