The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024526.2 | NP_001019697.2 | 14 - 15 | 1843 | 95 |
NM_001080948.2 | NP_001074417.1 | 14 - 15 | 1856 | 95 | |
NM_001284521.1 | NP_001271450.1 | 14 - 15 | 1830 | 95 | |
NM_001284522.1 | NP_001271451.1 | 14 - 15 | 1853 | 95 | |
NM_001284523.1 | NP_001271452.1 | 14 - 15 | 1846 | 95 | |
GenBank mRNA | AK049756.1 | - | 14 - 15 | 1770 | 95 |
AK139636.1 | - | 14 - 15 | 1851 | 95 | |
AK139909.1 | - | 14 - 15 | 1694 | 95 | |
AK165392.1 | - | 14 - 15 | 1789 | 95 |