Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Odf3b
Entrez Gene ID
70113
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
2010001J22Rik
Chromosome Location
Chr.15: 89377447 - 89379681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013022.1 NP_001013040.1 3 - 4 493 74
GenBank mRNA BC087962.1 - 3 - 4 493 74

View Full Product Details