The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198100.2 | NP_932768.2 | 8 - 9 | 1825 | 123 |
GenBank mRNA | BC057590.1 | - | 9 - 10 | 1783 | 123 |