Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scube3
Entrez Gene ID
268935
Gene Name
signal peptide, CUB domain, EGF-like 3
Gene Aliases
CEGF3, D030038I21Rik
Chromosome Location
Chr.17: 28142100 - 28174851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004366.1 NP_001004366.1 9 - 10 1072 70
GenBank mRNA AK083515.1 - 3 - 4 411 70
AY639609.1 - 9 - 10 1072 70
BC079849.1 - 7 - 8 821 70

View Full Product Details