The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030046.2 | NP_084322.2 | 2 - 3 | 341 | 70 |
GenBank mRNA | AK140224.1 | - | 2 - 3 | 342 | 70 |