The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205345.1 | NP_001192274.1 | 3 - 4 | 548 | 88 |
NM_001205346.1 | NP_001192275.1 | 3 - 4 | 548 | 88 | |
NM_194342.3 | NP_919323.2 | 3 - 4 | 548 | 88 | |
GenBank mRNA | AK046318.1 | - | 3 - 4 | 517 | 88 |
AK085196.1 | - | 3 - 4 | 517 | 88 | |
AK128958.1 | - | 3 - 4 | 542 | 88 | |
AK156246.1 | - | 3 - 4 | 549 | 88 | |
AK172996.1 | - | 3 - 4 | 373 | 88 | |
AY682987.1 | - | 2 - 3 | 314 | 88 | |
BC098208.1 | - | 3 - 4 | 468 | 88 |