Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl30
Entrez Gene ID
70788
Gene Name
kelch-like 30
Gene Aliases
4631423F02Rik
Chromosome Location
Chr.1: 91351073 - 91362404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027551.2 NP_081827.1 5 - 6 1294 78
GenBank mRNA AK014532.1 - 1 - 2 161 78
AK086025.1 - 5 - 6 1295 78
AK146580.1 - 5 - 6 1351 78
AK148398.1 - 5 - 6 1303 78
BC115428.1 - 4 - 5 1237 78
BC115429.1 - 4 - 5 1237 78

View Full Product Details