Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhl30
Entrez Gene ID
70788
Gene Name
kelch-like 30
Gene Aliases
4631423F02Rik
Chromosome Location
Chr.1: 91351073 - 91362404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027551.2 NP_081827.1 6 - 7 1482 82
GenBank mRNA AK014532.1 - 2 - 3 349 82
AK086025.1 - 6 - 7 1483 82
AK146580.1 - 6 - 7 1539 82
AK148398.1 - 6 - 7 1491 82
BC115428.1 - 5 - 6 1425 82
BC115429.1 - 5 - 6 1425 82

View Full Product Details