The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163945.1 | NP_001157417.1 | 9 - 10 | 1227 | 66 |
NM_025425.4 | NP_079701.1 | 6 - 7 | 894 | 66 | |
GenBank mRNA | AK002393.1 | - | 6 - 7 | 771 | 66 |
AK004280.1 | - | 6 - 7 | 895 | 66 | |
BC037498.1 | - | 6 - 7 | 652 | 66 | |
BC085243.1 | - | 6 - 7 | 795 | 66 | |
BC100562.1 | - | 4 - 5 | 492 | 66 | |
BC145876.1 | - | 6 - 7 | 720 | 66 | |
BC145878.1 | - | 6 - 7 | 720 | 66 |