The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012402.1 | NP_001012402.1 | 1 - 2 | 495 | 93 |
GenBank mRNA | AY574375.1 | - | 1 - 2 | 495 | 93 |
BC132520.1 | - | 1 - 2 | 482 | 93 | |
BC138814.1 | - | 1 - 2 | 482 | 93 |