Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnps1
Entrez Gene ID
19826
Gene Name
ribonucleic acid binding protein S1
Gene Aliases
-
Chromosome Location
Chr.17: 24414675 - 24425898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009070.2 NP_033096.2 2 - 3 177 114

View Full Product Details