Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fahd1
Entrez Gene ID
68636
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
1110025H10Rik, AV002085
Chromosome Location
Chr.17: 24848896 - 24850302 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023480.2 NP_075969.1 1 - 1 1317 80
GenBank mRNA AB041600.1 - 1 - 1 1151 80
AK044920.1 - 1 - 1 1116 80
AK141973.1 - 1 - 1 1317 80
AK157292.1 - 1 - 1 1316 80

View Full Product Details