The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008743.2 | NP_032769.2 | 5 - 6 | 851 | 87 |
GenBank mRNA | AB006812.1 | - | 5 - 6 | 797 | 87 |
AK033701.1 | - | 5 - 6 | 1859 | 87 | |
AV377313.2 | - | 3 - 4 | 439 | 87 | |
BC138852.1 | - | 5 - 6 | 814 | 87 | |
BC138855.1 | - | 5 - 6 | 814 | 87 | |
BC145443.1 | - | 5 - 6 | 723 | 87 | |
Y09688.1 | - | 5 - 6 | 823 | 87 |