The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026684.2 | NP_080960.1 | 3 - 4 | 479 | 124 |
GenBank mRNA | BC031664.1 | - | 3 - 4 | 479 | 124 |