Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spsb3
Entrez Gene ID
79043
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
Chromosome Location
Chr.17: 24886349 - 24892147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163750.1 NP_001157222.1 6 - 7 1162 69
NM_001163751.1 NP_001157223.1 6 - 7 817 69
NM_027141.2 NP_081417.2 6 - 7 838 69
GenBank mRNA AK082768.1 - 6 - 7 761 69
AK156579.1 - 5 - 6 1175 69
AK167474.1 - 5 - 6 1155 69
AK176934.1 - 2 - 3 177 69
AK191468.1 - 2 - 3 178 69
AK208055.1 - 2 - 3 177 69
AK210660.1 - 2 - 3 177 69
AK220234.1 - 6 - 7 1100 69
BC026523.1 - 6 - 7 1503 69
BC072556.1 - 6 - 7 1123 69

View Full Product Details