Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Josd1
Entrez Gene ID
74158
Gene Name
Josephin domain containing 1
Gene Aliases
1300006C06Rik, mKIAA0063
Chromosome Location
Chr.15: 79674250 - 79687872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028792.3 NP_083068.1 3 - 4 1460 95
GenBank mRNA AK004913.1 - 3 - 4 1409 95
AK154799.1 - 3 - 4 1509 95
AK190593.1 - 1 - 2 188 95
AK192247.1 - 1 - 2 188 95
AK196858.1 - 1 - 2 188 95
AK197502.1 - 1 - 2 188 95
AK197953.1 - 1 - 2 188 95
AK202351.1 - 1 - 2 189 95
AK206589.1 - 1 - 2 188 95
AK206648.1 - 1 - 2 188 95
AK208563.1 - 1 - 2 188 95
AK214040.1 - 2 - 3 324 95
AK218856.1 - 1 - 2 188 95
BC006928.1 - 3 - 4 608 95
BC086769.1 - 3 - 4 580 95

View Full Product Details