Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amhr2
Entrez Gene ID
110542
Gene Name
anti-Mullerian hormone type 2 receptor
Gene Aliases
Misiir, Misrii, Mrii
Chromosome Location
Chr.15: 102445028 - 102454640 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144547.2 NP_653130.2 5 - 6 717 81
GenBank mRNA AF503863.1 - 5 - 6 664 81
BC119571.1 - 5 - 6 648 81
BC119572.2 - 6 - 7 81

View Full Product Details