Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 14 - 15 2389 63
NM_001286041.1 NP_001272970.1 13 - 14 2326 63
NM_181413.4 NP_852078.1 14 - 15 2389 63
GenBank mRNA AK080878.1 - 3 - 4 1024 63
AK129094.1 - 13 - 14 2260 63
AK147273.1 - 14 - 15 2324 63
AK160806.1 - 1 - 2 63 63
AK169828.1 - 3 - 4 335 63
BC010633.1 - 2 - 3 272 63
BC050847.1 - 14 - 15 2355 63
BC057331.1 - 2 - 3 533 63

View Full Product Details