Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 19 - 20 3046 70
NM_001286041.1 NP_001272970.1 18 - 19 2983 70
NM_181413.4 NP_852078.1 19 - 20 3046 70
GenBank mRNA AK080878.1 - 8 - 9 1681 70
AK129094.1 - 18 - 19 2917 70
AK147273.1 - 19 - 20 2981 70
AK160806.1 - 6 - 7 720 70
AK169828.1 - 8 - 9 992 70
BC010633.1 - 7 - 8 929 70
BC050847.1 - 19 - 20 3012 70
BC057331.1 - 7 - 8 1190 70

View Full Product Details