Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 8 - 9 1216 61
NM_001286041.1 NP_001272970.1 7 - 8 1153 61
NM_181413.4 NP_852078.1 8 - 9 1216 61
GenBank mRNA AK129094.1 - 7 - 8 1087 61
AK147273.1 - 8 - 9 1151 61
BC050847.1 - 8 - 9 1182 61

View Full Product Details