Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbln1
Entrez Gene ID
14114
Gene Name
fibulin 1
Gene Aliases
-
Chromosome Location
Chr.15: 85205944 - 85286294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010180.2 NP_034310.2 7 - 8 918 112
GenBank mRNA AK035388.1 - 7 - 8 871 112
AK083573.1 - 7 - 8 921 112
AK085989.1 - 7 - 8 861 112
AK086451.1 - 7 - 8 919 112
AK137511.1 - 7 - 8 861 112
BC007140.1 - 7 - 8 862 112
EU543224.1 - 7 - 8 856 112
X70854.1 - 7 - 8 915 112

View Full Product Details