The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005868.2 | NP_001005868.1 | 1 - 2 | 336 | 116 |
NM_001289473.1 | NP_001276402.1 | 1 - 2 | 336 | 116 | |
NM_001289474.1 | NP_001276403.1 | 1 - 2 | 336 | 116 | |
NM_001289475.1 | NP_001276404.1 | 1 - 2 | 336 | 116 | |
GenBank mRNA | AK169529.1 | - | 1 - 2 | 265 | 116 |
BC141178.1 | - | 1 - 2 | 45 | 116 | |
BC141188.1 | - | 1 - 2 | 45 | 116 | |
BC145498.1 | - | 1 - 2 | 45 | 116 | |
BC145509.1 | - | 1 - 2 | 45 | 116 | |
BM933013.1 | - | 2 - 3 | 116 | ||
BY745732.1 | - | 1 - 2 | 265 | 116 |