The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177857.1 | NP_808525.2 | 5 - 6 | 1490 | 148 |
GenBank mRNA | AK044389.1 | - | 4 - 5 | 1866 | 148 |
BC060666.1 | - | 5 - 6 | 1490 | 148 |