The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177857.1 | NP_808525.2 | 10 - 11 | 1991 | 59 |
GenBank mRNA | AK044389.1 | - | 9 - 10 | 2367 | 59 |
AK137637.1 | - | 5 - 6 | 598 | 59 | |
AK172232.1 | - | 6 - 7 | 713 | 59 | |
BC046440.1 | - | 5 - 7 | 459 | 59 | |
BC060666.1 | - | 10 - 11 | 1991 | 59 | |
BC066182.1 | - | 3 - 4 | 237 | 59 |