The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039347.1 | NP_001034436.1 | 5 - 6 | 1921 | 78 |
GenBank mRNA | AF107782.1 | - | 4 - 5 | 1456 | 78 |
AK147574.1 | - | 5 - 6 | 1832 | 78 |