The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008291.3 | NP_032317.2 | 1 - 2 | 219 | 91 |
GenBank mRNA | BB621153.1 | - | 1 - 2 | 219 | 91 |
U66827.1 | - | 1 - 2 | 204 | 91 |