The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028480.2 | NP_082756.1 | 7 - 8 | 2398 | 126 |
GenBank mRNA | AK145976.1 | - | 7 - 8 | 2399 | 126 |
AK146102.1 | - | 7 - 8 | 2400 | 126 | |
BC100349.1 | - | 7 - 8 | 2376 | 126 |