The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025292.6 | NP_079568.1 | 3 - 4 | 429 | 74 |
GenBank mRNA | AK008054.1 | - | 3 - 4 | 412 | 74 |
AK013474.1 | - | 3 - 4 | 391 | 74 | |
AK030379.1 | - | 2 - 3 | 286 | 74 | |
AK033514.1 | - | 3 - 4 | 430 | 74 | |
AK078422.1 | - | 2 - 3 | 286 | 74 | |
AK163362.1 | - | 3 - 4 | 397 | 74 | |
AY138960.1 | - | 3 - 4 | 369 | 74 | |
BC027433.1 | - | 3 - 4 | 391 | 74 |