Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc78
Entrez Gene ID
381077
Gene Name
coiled-coil domain containing 78
Gene Aliases
Gm938, Gm950
Chromosome Location
Chr.17: 25785619 - 25790513 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165929.1 NP_001159401.1 11 - 12 1150 77
GenBank mRNA AK140238.1 - 10 - 11 1618 77

View Full Product Details